Learn More
Drummond™ Original Portable Pipet-Aid™ Pipet Controller
Convenient alternative to mouth pipetting in the laboratory.
Brand:  Drummond™ 07897-32
726.75 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 11714819
  • 765.00 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easy-to-use volume control mechanism permits precise fluid intake and delivery
  • Check valve protects against accidental fluid intake into the handle
  • Can be used with glass or plastic pipets
  • Autoclavable components
Order Info
Contact your Fisher Scientific Customer Service Representative for available replacement parts.
Certifications
CE, Euro version
Specifications
Specifications
Autoclavable
Laboratory pipetting
Pipet Controller, TC Nosepiece
Portable Filler/Dispenser
Glass or plastic pipets
220 V