missing translation for 'onlineSavingsMsg'
Learn More
Grant Instruments™ Dry Block Heater/Cooler Product Code.: 11441018
Estimated Shipment: 03-07-2025
to see stock.
Add to basket

Grant Instruments™ Dry Block Heater/Cooler
GREENER_CHOICE

Product Code. 11441018
Holds up to 32 microtubes
Click to view available options
Holds:
Holds up to 32 microtubes
This item is not returnable. View return policy

Product Code. 11441018

Brand: Grant Instruments™ PCH1 12V

Estimated Shipment: 03-07-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Maintains the set temperature on the aluminum block with special sockets for tubes. Grant Instruments™ Dry Block Heater/Cooler works best within temperatures ranging from -10 to 100°C.

For use with laboratory applications requiring cooling and heating in microtubes within the temperature setting.
  • Switches easily between cooling and heating modes
  • Advantageous when working with the microquantities of reagents used in microtubes
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
    • Recyclable
TRUSTED_SUSTAINABILITY

Specifications

Holds Holds up to 32 microtubes
Temperature Range (Metric) -10°C to +100°C
Type Dry Bath Block
Temperature Stability ±0.1°C
Temperature Uniformity ±0.5°C
Timer 1 min. to 96 hr.
Warranty 2 Years
Dimensions (L x W x H) 260 x 240 x 165 mm
Length (Metric) 260 mm
Width (Metric) 240 mm
Height (Metric) 165 mm
Weight (Metric) 3.6 kg
Voltage 120/230 V
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Grant Instruments™ Dry Block Heater/Cooler >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.