Learn More
3M™ Dry-Sponge BP133ES
Biocide-free cellulose sponge that comes with a printed sample bag.
Brand:  3M™ 7000042938
Encompass
Product Code. 17238530
  • 221.85 € / Pack of 100
Explore more special offers
Description
Description
  • 1.5 x 3 in. dry sponge can be used with a wide range buffers and enrichment media and in dry-only environments.
  • Simplified packaging convenience minimizes handling and preparation and reduces waste.
  • Biocide-free sponge does not affect organism viability.
  • 5 x 9 in. printable sample bag included for safe, convenient transportation to lab.
  • Decontaminated through gamma irradiation to maintain aseptic conditions.
Specifications
Specifications
Dry Sponge
1.5 x 3 in.
100/Cs.