Learn More
3M™ Dry-Sponge with Glove BP237SPG
Biocide-free cellulose sponge that includes two polyethylene gloves and a sample bag.
Brand:  3M™ 7000042939
Encompass
Product Code. 17248530
  • POA / Pack of 100
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • 1.5 x 3 in. dry sponge can be used with a wide range buffers and enrichment media and in dry-only environments.
  • Simplified packaging convenience, including sponge, sample bag and gloves, minimizes handling and preparation and reduces waste.
  • 5 x 9 in. sample bag with 18 oz./0.53 L capacity and included for safe handling and convenient transportation to lab.
  • Biocide-free sponge does not affect organism viability.
  • Decontaminated through gamma irradiation to maintain aseptic conditions.
Specifications
Specifications
Dry Sponge
1.5 x 3 in.
100/Cs.