Learn More
Lenz Laborglasintrumente Drying Tubes, Bent, (Calcium Chloride Tubes)
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
10.00 € - 11.00 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10356261
|
Lenz Laborglasintrumente
05333013 |
150 mm |
11.00 €
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
10010221
|
Lenz Laborglasintrumente
05333010 |
100 mm |
10.00 €
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
Description
- Drying tubes acc. to DIN 12610, bent
- With ball and hose connection
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) |