missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DrySpot™Staphytect Plus™Latex Agglutination Test

Rapidly identify and differentiate Staphylococcus aureus by detecting clumping factor, Protein A, and certain capsular polysaccharides.

Brand:  Thermo Scientific™ DR0100M

Product Code. 11439868

  • 250.00 € / 120 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Rapidly identify and differentiate Staphylococcus aureus with Thermo Scientific™ DrySpot™ Staphytect Plus™ Latex Agglutination Test.  DrySpot Staphytect Plus detects clumping factor, Protein A, and certain capsular polysaccharides found in methicillin resistant S.s aureus (MRSA) to differentiate S.s aureus from staphylococci that do not possess these properties. Test cards contain pre-dispensed reagents that are dried onto the reaction areas for test convenience, extended shelf-life when compared to wet reagents, and room temperature storage.

DrySpot Staphytect Plus latex agglutination test uses blue latex particles, which are coated with porcine fibrinogen and rabbit IgG, then pre-dried onto reaction cards for the detection of clumping factor, Protein A, and capsular polysaccharides of S. aureus.
  • Long shelf-life: two years from the date of manufacture
  • Flexible storage: can be stored in a fridge or at room temperature (2-25°C)
  • No dispensing errors: reagents are pre-dried onto reaction cards
  • Easy-to-read blue latex
  • Rapid results: within 20 seconds
  • Excellent sensitivity: detects three properties of S. aureus for wider strain identification
Traditionally, differentiation between coagulase-positive and coagulase-negative staphylococci has been performed either with the tube coagulase test that detects extracellular staphylocoagulase or the slide coagulase test that detects the clumping factor (bound coagulase) present on the bacterial cell surface. It has been reported that approximately 97% of human strains of S. aureus possess both bound coagulase and extracelluar staphylocoagulase. Protein A is found on the cell surface of about 95% of human strains of S. aureus and has the ability to bind the Fc portion of immunoglobulin G (IgG)1.
Certain methicillin-resistant strains of S. aureus (MRSA) may express undetectable levels of clumping factor and Protein A2,3,4. It has been shown, however, that these strains possess capsular polysaccharide5. The capsule can mask both Protein A and the clumping factor, thereby, preventing agglutination.
DrySpot Staphytect Plus uses latex particles coated with porcine fibrinogen and rabbit IgG, including specific polyclonal antibodies raised against capsular polysaccharides of S. aureus6,7. When the dried reagents are mixed with a suspension of S. aureus colonies, rapid agglutination occurs through the reaction between (i) fibrinogen and clumping factor, (ii) the Fc portion of IgG and Protein A, (iii) specific IgG and capsular polysaccharide.
Agglutination may also occur with other species which possess clumping factor or Protein A, such as Staphylococcus hyicus and Staphylococcus intermedius. If neither clumping factor, Protein A, or specific capsular polysaccharides are present, agglutination will not occur, and the result will be regarded as negative. The most frequent coagulase and Protein A negative isolates of staphylococci are Staphylococcus epidermidis.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2°C to 25°C
DrySpot Staphytect Plus Kit
Pouches containing reaction cards with test and control latex, plastic pouch clip
120 Tests/Kit
Positive
Staphylococcus aureus Test Kit
Latex
Staphylococcus aureus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.