Learn More
Fisherbrand™ Anodized Dual Aluminum Blocks
Useful for a variety of applications in molecular biology, histology, clinical, environmental and industrial. Fisherbrand™ Anodized Dual Aluminum Blocks are for use with Fisherbrand Digital Block Heaters.
Brand: Fisherbrand™ D1296
This item is not returnable.
View return policy
Description
- Excellent heat transfer
- Anodized to resist thermal shock
- For holding test tubes for digital heaters
- Available to fit a range of tube counts and sizes
Specifications
Micro plate or 4 Slides. | |
D1196, 96-Well | |
1 CT |