Learn More
Fisherbrand™ Dual LCD Display Thermo-Hygrometer
Brand:  Fisherbrand™ 30.5002
Product Code. 11945148
  • 32.30 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Specifications
Thermohygrometer
0.01
Dual LCD
110 mm
2-AAA Batteries
10 to 99%
-10°C to +60°C
110 mm
1.5 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Dual LCD Display Thermo-Hygrometer > Dimensions: 110L x 20D x 110mmH; Temperature Range: -10 to 60deg.C