Learn More
PureAire Monitoring Systems, Inc. Dual O2/CO2 (0-25% O2, 0-50,000ppm CO2) with 10+ Year Sensors
Perfect for your facility if you use nitrogen, helium, and/or argon gases. The monitor is also suited for use in confined spaces such as cryogenic facilities. Sensors lasting 10 years and more.
Brand:  PureAire Monitoring Systems, Inc. 99144
Product Code. 16228831
  • 3735.00 € / Each
Explore more special offers
Description
Description
PureAire Monitoring System’s Dual O2/CO2 Monitor provides continuous monitoring of oxygen and carbon dioxide. This monitor is ideal for use in spaces where low oxygen concentrations could pose a health hazard, from nitrogen gas storage sites to CO2 beverage gas distribution areas and more.
  • No maintenance zirconium O2 cell and long life CO2 sensor
  • No calibration required for oxygen Self-calibrating CO2 sensor
  • Local backlit concentration display
  • Built-in audible horn
  • Built-in visual LED alarm indicators
  • Alarm level 1 and 2 for O2 and CO2
  • No drift due to environmental or temperature changes
  • Plug-in the wall power supply included- 24V, 220V
What is a Dual O2/CO2 Monitor?
  • A Dual O2/CO2 Monitor is a stand-alone system ideal for the continuous monitoring of nitrogen or cryogenics near freezers, confined spaces, and other locations where low oxygen levels may pose a hazard to personnel.
Why choose PureAire?
  • As an industry leader in oxygen monitoring and gas detection, PureAire is dedicated to the safety of our clients, and the customers they serve, by manufacturing monitors that are long-lasting, innovative, and reliable.
What gases can a Dual O2/CO2 Monitor be used for?
  • PureAire’s oxygen monitors can be used for CO2, nitrogen, argon, and helium gases.
Installation is easy. Plug-in ready.
  • The monitor will arrive ready to work right out of the box. Simply secure the unit on the wall 1-2 meters from the gas source, plug it in, and power it on. The monitor will begin sampling oxygen levels after a 4 minute startup period. Levels are displayed clearly on the screen for easy access and visibility.  
Certifications
CE Marked
Services
2 years
Specifications
Specifications
CE
76 mm
3.15 in.
0-95% RH
-40 to +55°C
2 Year after ETD
0.5 kg
165 mm
3 in.
3 x 6.5 x 3.15 in.
80 mm
-40°F to +134°F
24V
1.1 lb.
6.5 in.