Learn More
Thermo Scientific™ Dual QuantStudio System Integration Kit for the Orbitor™ RS2 Microplate Mover and QuantStudio™ Real-time PCR Systems
Designed for use with the Orbitor RS2 Microplate Mover and identical QuantStudio Systems, enabling real-time PCR in automated environments for increased sample throughput
Brand:  Thermo Scientific™ A43779
Product Code. 17142504
  • 9740.00 € / Each
Explore more special offers
Description
Description
  • This Dual QuantStudio System Integration Kit is designed for use with the Orbitor RS2 Microplate Mover and identical QuantStudio Systems, enabling real-time PCR in automated environments for increased sample throughput.
  • This integration kit includes a location bracket, splitter, and Thermo Scientific Momentum Workflow Software for the second QuantStudio integration.
  • Couple the Dual QuantStudio Integration Kit with the rbitor RS2 Microplate Mover for QuantStudio Systems and one of the two plate storage and handling options (Sequential Access or Random Access ) to complete your automation bundle.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
QuantStudio 7 Pro, QuantStudio 7 Flex, or QuantStudio 12 K Flex Real-Time PCR Systems and The Orbitor RS2 Microplate Mover
Real Time PCR (qPCR)
1 kit
Integration Kit
Room Temperature