Learn More
Hamilton™ Dual Small Hub RN Coupler
Small Hub Removable (RN) needles can be used with removable (RN)-style syringes that are 100 μL and smaller. Standard needle length is 2 in. / 51 mm. Removable (RN) needles are available in convenient six packs. Custom lengths and point styles are available.
Brand:  Hamilton™ 55752-01
Product Code. 12366239
  • 78.00 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Dual Small Hub RN Coupler