Learn More
Corning™ Dulbecco's Phosphate Buffered Saline
BIOPHARMA
For use alone or combined with other agents for washing tissues and cells
105.71 € - 498.62 €
Specifications
Content And Storage 2° to 30°C
Product Type DPBS
View More Specs
Products 3
Product Code Brand Capacity Price Quantity & Availability  
Product Code Brand Capacity Price Quantity & Availability  
15343571
BIOPHARMA
View Documents
Corning™
20-031-CV
500 mL
105.71 €
Pack of 6
15353631
BIOPHARMA
View Documents
Corning™
21-031-LB
20,000 mL
498.62 €
20 L
15363631
BIOPHARMA
View Documents
Corning™
21-031-LX
10,000 mL
303.78 €
10 L
Description
Description
The Corning range of buffered salt solutions for cell culture are made to a physiological pH and salt concentration (after appropriate dilution). They are used alone or in combination with other agents for washing tissues and cells. Buffered salt solutions most commonly include sodium, potassium, calcium, magnesium, and chloride. They provide the cells with water and inorganic ions, while maintaining a physiological pH and osmotic pressure. Although there have been many modifications to the original formulae in efforts to produce fully-defined media, discrete salt solutions still play an important role in tissue culture research. The basic function of these solutions, i. e. , to maintain the pH and osmotic balance in the medium and to provide the cells with water and essential inorganic ions, are as valuable today as when they were first developed a century ago.
Specifications
Specifications
2° to 30°C
DPBS
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 18 Months