missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Dunlop™ Hygrade Boots

Product Code. 12054475
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12054475

Brand: Euro Protection™ 9HYGR46

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
  • Suitable for food industry.
  • PVC upper.
  • Nylon lining.
  • PVC / Nitrile outsole.

Specifications

Color White, Green
For Use With (Application) Food industry
Liner Material Nylon
Material PVC, Nitrile, Nylon
Size 46
Sole Material PVC, Nitrile
Weight (Metric) 1.075 kg per boot in size 44
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.