missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Dunlop™ Selenium Boots

35.00 €

Specifications

Certifications/Compliance CE Certified
Color Green, Black, White
Footbed Material PVC
For Use With (Application) For working purpose
Liner Material Polyester
View More Specs

Products 1
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12094525
View Documents
Euro Protection™
9SELE40
40
35.00 €
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

CE Certified
PVC
Polyester
Black color, anti-slip sole
Non-safety
Yes
Green, Black, White
For working purpose
PVC
PVC
Water, hydrocarbures
Boot
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.