missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Duomax 1030 (5° Tilt Angle)

Platform shaker that keeps containers safely fixed on an anti-slip rubber mat, even at high speeds

Brand:  Heidolph™ 543-32205-06

Product Code. 17333030

  • 1910.00 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Containers are safely fixed on the anti-slip rubber mat even at high speeds. The speed is variably adjustable between 2 and 50 rpm. The shaker can be operated in continuous operation or with a timer (max. 120 minutes).
  • A tilt angle of 5° is recommended for applications for which a gentle movement amplitude is suitable. If the application requires stronger shaking, the model can also be obtained with an angle of 10°.
  • The Duomax 1030 can be integrated into the modular incubator model.
Specifications

Specifications

230 V
UK Plug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.