Learn More
Macherey-Nagel™ Duotest™ pH Paper Roll
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Rolls give the user the best possible accuracy in pH measurement.
Brand: Macherey-Nagel™ 90302
This item is not returnable.
View return policy
Description
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Rolls give the user the best possible accuracy in pH measurement.
- Provides rapid and reliable results
- Available for a range of pH gradations
- Shows two different colours for each pH-value at intervals of 0.3 to 1 pH unit
- Allows more accurate reading and good estimation of intermediate values
Specifications
1 to 4.3 | |
1.0, 1.3, 1.6, 1.9, 2.2, 2.5, 2.8, 3.1, 3.4, 3.7, 4.0, 4.3 | |
pH Paper | |
10 mm |
Roll | |
1 Each | |
5 m |
Product Content Correction
Product Title
Macherey-Nagel™ Duotest™ pH Paper Roll > pH Range: 1 to 4.3
Spot an opportunity for improvement?Share a Content Correction