missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keystone Cleanroom Products DuPont™ Tyvek™ Autoclave Pouch with Steam Indicator Dot
Manufactured in a ISO6 cleanroom to ensure cleanliness.
344.00 € - 872.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Capacity (English) | Application Dependent |
Certifications/Compliance | Certificate of Conformance (COC) |
Closure Type | Heat Seal |
Color | White |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
17708267
|
Keystone Cleanroom Products
COV-MGTVK-UB-6X8 |
8 in. x 6 in. |
472.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17748267
|
Keystone Cleanroom Products
COV-MGTVK-UB-8X9 |
9 in. x 8 in. |
490.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17718267
|
Keystone Cleanroom Products
COV-MGTVK-UB-8X12 |
12 x 8 in. |
444.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17738237
|
Keystone Cleanroom Products
COV-MGTVK-UB-10X12 |
12 in. x 10 in. |
509.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17728267
|
Keystone Cleanroom Products
COV-MGTVK-UB-8X14 |
14 in. x 8 in. |
518.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17778257
|
Keystone Cleanroom Products
COV-MGTVK-UB-6X16 |
16 in. x 6 in. |
539.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17768237
|
Keystone Cleanroom Products
COV-MGTVK-UB-12X18 |
18 in. x 12 in. |
595.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17748237
|
Keystone Cleanroom Products
COV-MGTVK-UB-10X20 |
20 in. x 10 in. |
585.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17768247
|
Keystone Cleanroom Products
COV-MGTVK-UB-20X20 |
20 in. x 20 in. |
518.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17788257
|
Keystone Cleanroom Products
COV-MGTVK-UB-6X22 |
22 in. x 6 in. |
344.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17788237
|
Keystone Cleanroom Products
COV-MGTVK-UB-14X22 |
22 in. x 14 in. |
708.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17718257
|
Keystone Cleanroom Products
COV-MGTVK-UB-30X22 |
22 in. x 30 in. |
621.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17728247
|
Keystone Cleanroom Products
COV-MGTVK-UB-18X24 |
24 in. x 18 in. |
500.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17768257
|
Keystone Cleanroom Products
COV-MGTVK-UB-5X25 |
25 in. x 5 in. |
539.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17758237
|
Keystone Cleanroom Products
COV-MGTVK-UB-10X25 |
25 in. x 10 in. |
367.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17798237
|
Keystone Cleanroom Products
COV-MGTVK-UB-14X28 |
28 in. x 14 in. |
774.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17708257
|
Keystone Cleanroom Products
COV-MGTVK-UB-26X28 |
28 in. x 26 in. |
641.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17758257
|
Keystone Cleanroom Products
COV-MGTVK-UB-48X28 |
28 in. x 48 in. |
872.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17738267
|
Keystone Cleanroom Products
COV-MGTVK-UB-8X30 |
30 in. x 8 in. |
626.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17738247
|
Keystone Cleanroom Products
COV-MGTVK-UB-18X30 |
30 in. x 18 in. |
574.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17778237
|
Keystone Cleanroom Products
COV-MGTVK-UB-12X32 |
32 in. x 12 in. |
718.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17778247
|
Keystone Cleanroom Products
COV-MGTVK-UB-20X32 |
32 in. x 20 in. |
610.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17798257
|
Keystone Cleanroom Products
COV-MGTVK-UB-6X34 |
34 in. x 6 in. |
595.00 €
Pack of 100 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17728257
|
Keystone Cleanroom Products
COV-MGTVK-UB-34X34 |
34 in. x 34 in. |
749.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17748257
|
Keystone Cleanroom Products
COV-MGTVK-UB-3X36 |
36 in. x 3 in. |
385.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17708247
|
Keystone Cleanroom Products
COV-MGTVK-UB-16X36 |
36 in. x 16 in. |
585.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17798247
|
Keystone Cleanroom Products
COV-MGTVK-UB-25X37 |
37 in. x 25 in. |
713.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17748247
|
Keystone Cleanroom Products
COV-MGTVK-UB-18X40 |
40 in. x 18 in. |
641.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17738257
|
Keystone Cleanroom Products
COV-MGTVK-UB-38X40 |
40 in. x 34 in. |
872.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17718247
|
Keystone Cleanroom Products
COV-MGTVK-UB-16X44 |
44 in. x 16 in. |
636.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17788247
|
Keystone Cleanroom Products
COV-MGTVK-UB-24X44 |
44 in. x 24 in. |
764.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17758247
|
Keystone Cleanroom Products
COV-MGTVK-UB-18X68 |
68 in. x 18 in. |
754.00 €
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
Description
- Designed for steam sterilization of pharmaceutical production equipment and components
- 1073B Tyvek™ construction allows for optimal steam penetration during sterilization
- Excellent impact resistance
- Consistently strong factory seals
- Exceptional microbial barrier
- Resists tearing and punctures from sharp corners and heavy machine parts
- Processed and packaged in a controlled cleanroom environment
- Lot traceable
Specifications
Autoclavable | |
Certificate of Conformance (COC) | |
White | |
Steam Sterilization | |
Tyvek | |
Steam |
Application Dependent | |
Heat Seal | |
Single-use | |
Pharmaceutical Equipment Change Parts/Components (Stoppers, Seals, etc.) | |
1615 g/m2/24 hr |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title