missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products DuPont™ Tyvek™ Autoclave Pouch with Steam Indicator Dot
GREENER_CHOICE

Autoclave Pouch with Steam Indicator Dot - Manufactured in a ISO6 cleanroom to ensure cleanliness.

Brand:  Keystone Cleanroom Products COVMGTVKUB28X42

 View more versions of this product

Product Code. 17831081

  • 1050.00 € / Pack of 50
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Designed for steam sterilization of pharmaceutical production equipment and components
  • 1073B Tyvek™ construction allows for optimal steam penetration during sterilization
  • Excellent impact resistance
  • Consistently strong factory seals
  • Exceptional microbial barrier
  • Resists tearing and punctures from sharp corners and heavy machine parts
  • Lot traceable
  • Certificates of Conformance sent with shipment
Specifications

Specifications

Autoclavable
31.6 L
Heat Seal
Single-use
Steam Indicator
Non-sterile
Yes
Steam
8 mil
42 in.
28 in.
8.35 gal.
Certificate of Conformance
White
Steam Sterilization of Pharmaceutical Equipment
Tyvek
0.200 mm
50
121°C to 125°C
28 x 42 in.
107 cm
71.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.