Learn More
H-B Instrument™ Durac™ Barometer
Recalibrateable unit
112.62 €
Description
- Features black lines and numbers
- Hole present for wall mounting
- Adjust automatically once set
- Designed to work in altitudes below 6562 ft. (2000m)
Specifications
Barometer |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
H-B Instrument™ Durac™ Barometer