Learn More
H-B Instrument™ DURAC™ Digital Stopwatches
Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) instructions, these electronic stopwatches include a calendar and a clock.
Brand:  H-B Instrument™ B61700-5300
 View more versions of this product
Product Code. 17673175
  • 54.50 € / Each
Explore more special offers
Description
Description
  • Features Certificate of Calibration and individual serial number
  • Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
  • B61700-5200 stores 30 lap times and calculates lap time average, allows for 99 split lap times, and has a 10/320SEC pacer
  • B61700-5300 will store 1 lap time and includes an alarm
  • Requires one CR2032 battery (included)
Specifications
Specifications
Stopwatch
0.01 sec
With Clock, Alarm and Calendar
63 mm
80 mm
Chronograph: 1/100 sec
0.001
2.5 in.
3.1 in.