missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- 10.01 pH value
- Immune to micro-organisms
- Expiration date on the bottle
- First class certificate with traceability to international standards
Specifications
Specifications
Color | Yellow |
pH | 10.01 |
Physical Form | Liquid |
Quantity | 500 mL |
Solution Type | Buffer |
Solubility Information | Water Soluble |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuraCal™ Buffer pH 10.01, Hamilton™ >
Spot an opportunity for improvement?Share a Content Correction