Learn More
DuraCal™ 10.01 Value pH Buffer, Hamilton™
Click to view available options
Quantity:
500 mL
Description
- 10.01 pH value
- Immune to micro-organisms
- Expiration date on the bottle
- First class certificate with traceability to international standards
Specifications
Specifications
pH | 10.01 |
Quantity | 500 mL |