missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuraCal™ 9.21 Value pH Buffer, Hamilton™
Click to view available options
Quantity:
500 mL
Description
- 9.21 pH value
- Immune to micro-organisms
- Expiration date on the bottle
- First class certificate with traceability to international standards
Specifications
Specifications
pH | 9.21 |
Quantity | 500 mL |
Safety and Handling
Signal Word
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction