Learn More
DWK Life Sciences DURAN™ Adapter, standard ground joint
The transition adapter connects two glass components with different standard ground joints.
Brand:  DWK Life Sciences 241144405
 View more versions of this product
Product Code. 10151451
  • 31.35 € / Each
Estimated Shipment: 28-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Transition adapters or reducers are glass components with two standard ground joints of different sizes. With the help of these adapters, glass components with different standard ground joints can be connected to each other.
  • Tight closure due to standard ground joint
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Ea.
Connection of two glass components
Reduction Joint, Non-DIN Size
Socket NS 29/32, Cone NS 24/29