missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Standard Taper Socket Side Arm, Unground Neck, without Stopper
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
1000 mL
2000 mL
500 mL
Unit Size:
Each
Pack of 10
Description
Aspirator bottles, ground glass side-arm fro dispensing of liquids. Complying with ISO 4796, these aspirators have a ground socket side-arm and plain neck to accept rubber stoppers. Compatible stopcocks assemblies are available separately.
- Outlet facilitates attachment of ground jointed stopcock
- With plain unground neck for rubber stopper and bottom ground joint outlet
- Made of virtually inert, chemically resistant DURAN™ glass
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| For Use With (Application) | Storage and dispensing of liquids and reagents |
| Material | Borosilicate Glass (3.3) |
| No. per Pack | 10/Pk. |
| Neck Finish | Standard Taper Joint Bottom 19/26 |
| Diameter (Metric) | 86 mm |
| Autoclavable | Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction