missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Standard Taper Socket Side Arm, Unground Neck, without Stopper
Aspirator bottle with ground joint glass side-arm.
Brand: DWK Life Sciences 247015409
This item is not returnable.
View return policy
Description
Aspirator bottles, ground glass side-arm fro dispensing of liquids. Complying with ISO 4796, these aspirators have a ground socket side-arm and plain neck to accept rubber stoppers. Compatible stopcocks assemblies are available separately.
- Outlet facilitates attachment of ground jointed stopcock
- With plain unground neck for rubber stopper and bottom ground joint outlet
- Made of virtually inert, chemically resistant DURAN™ glass
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
1000 mL | |
Borosilicate Glass (3.3) | |
Standard Taper Joint Bottom 19/26 | |
107 mm |
Storage and dispensing of liquids and reagents | |
10/Pk. | |
Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction