Learn More
DWK Life Sciences DURAN™ Bell Jar, with glass knob top
Flat flange bell with knob and ground flange for mounting on a vacuum experimental plate (vacuum chamber).
Brand:  DWK Life Sciences 244606607
349.60 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10304961
  • 396.00 € / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
By default, tests are carried out at normal pressure. The conduct of tests under vacuum (negative pressure) can have significant effects on the reaction sequence. In order to be able to observe these effects in the rough and fine vacuum range, a vacuum experiment plate and a bell jar (also called air pump bell) are required.
  • The button makes it easy to raise the bell jar
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY
Specifications
Specifications
260 mm
Vacuum applications; Tests in vacuum chambers
Autoclavable
Borosilicate Glass (3.3)
1/Pk.