Learn More
DWK Life Sciences DURAN™ Blank for separating and dropping funnel, cylindrical with stopper bed for ISO 4800
Raw body for further processing as dropping funnel.
Brand:  DWK Life Sciences 210142404
 View more versions of this product
Product Code. 15231629
  • 150.00 € / Pack of 10
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DURAN™ dropping funnels are used, for example, for the metered supply of liquid to a reaction mixture.
  • Cylindrical with stopper bed for ISO 4800
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
42 mm
Borosilicate Glass (3.3)
Blank for further processing as dropping funnel; Phase separation
100 mL
Autoclavable
10/Pk.