Learn More
Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Angled
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Angled are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03314158
 View more versions of this product
Product Code. 10342751
  • 49.00 € / Each
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12394
  • Two side necks at 20° angles
  • With conical ground joints, featuring high taper accuracy and excellent tightness
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
500 mL
Borosilicate Glass, Duran Glass
14/23 NS Side Neck, 29/32 NS Center Neck
Round