Learn More
Hirschmann™ DURAN™ Burette Tubes with Blue Graduations
Burette assembly with burette tube and separate PTFE stopcock with PP tip. Excellent for titrating exact amounts of chemical solutions in the laboratory.
Brand:  Hirschmann™ 3071270
 View more versions of this product
Product Code. 13187959
  • 48.80 € / Pack of 2
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Short line
  • Blue graduation
Specifications
Specifications
Burettes tube DURAN™, blue graduation, for burette stopcock (890 10 76)
8 mm
500 mm
2/Pack
Blue
25 mL
Burette Stopcock
Buret Tube