Learn More
Hirschmann™ DURAN™ Burette Tubes with Blue Graduations
Burette assembly with burette tube and separate PTFE stopcock with PP tip. Excellent for titrating exact amounts of chemical solutions in the laboratory.
Brand: Hirschmann™ 3071275
This item is not returnable.
View return policy
Description
- Short line
- Blue graduation
Specifications
Burettes tube DURAN™, blue graduation, for burette stopcock (890 10 76) | |
50 mL | |
2/Pack | |
Buret Tube |
8 mm | |
500 mm | |
±0.075 mL |