Learn More
Hirschmann™ DURAN™ Class AS Micro Burettes Blue Graduations
Excellent for dispensing measured amounts of chemical solutions in the laboratory. Manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 3310152
 View more versions of this product
Product Code. 13148009
  • 443.00 € / Pack of 2
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Schellbach
  • Bang pattern
  • Main point ring graduation
  • With blue graduation
  • Acc. to DIN EN ISO 385
  • Support base
  • Conformity certified, with dated batch identification
Specifications
Specifications
Micro Burettes acc. to Bang, Class AS, conformity certified
DURAN™, burette and stopcock key
Bang, Main Point Ring Graduation, Schellbach Stripes, Socket
Blue
Glass
Class AS
5 mL
0.02 mL
Lateral stopcock, standard ground glass key
Lateral
±0.01 mL
DIN EN ISO 385