Learn More
DWK Life Sciences DURAN™ Crystallizing Dishes with Spout
An excellent option for crystallization, heating and cooling baths and microscale experiments in the laboratory.
72.22 €
Specifications
Autoclavable Autoclavable
View More Specs
Products 1
Product Code Brand Capacity (Metric) Diameter (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Price Quantity & Availability  
10759101
View Documents
DWK Life Sciences
213115904
2000 mL 190 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Acc. to DIN 12338
  • Transparent with labelling patch
  • Glass Type I/neutral glass as per USP, EP and JP
  • Autoclavable
  • Available in a range of capacities
Specifications
Specifications
Autoclavable