missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Dropping Funnel, cylindrical, with scale

The dropping funnel allows a dosed addition of solutions.

Brand:  DWK Life Sciences 241221704

99.75 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10439922

  • 111.00 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The dosed addition of solutions, reaction mixtures and/or chemicals is ideally carried out using a dropping funnel.

  • With standard ground stopcock and retaining device
  • The rate of supply can be adjusted
  • Cylindrical with scale
  • Tight closure due to standard ground joint
  • Fired-on, highly durable white print
  • Easy-to-read scale with graduation marks to estimate the content
TRUSTED_SUSTAINABILITY
Specifications

Specifications

9 mm
Boro 3.3, Polypropylene
Uniform and metered liquid supply to a reaction mixture; Measuring liquid amounts
1 Ea.
50 mL
Autoclavable
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers