Learn More
DWK Life Sciences DURAN™ Drying Tube, bent, with standard ground cone
Filled with a desiccant, the drying tube protects a reaction mixture in a flask from moisture.
Brand:  DWK Life Sciences 242620609
25.22 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10614182
  • 28.55 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The drying tube is usually an angled glass tube with a standard ground joint at one end. The other end is open. For filling the drying tube is first stuffed with a little cotton wool, then the drying agent is filled in and finally another piece of cotton wool prevents the drying agent from falling out.
  • Tight closure due to standard ground joint
  • Bent
  • With standard ground socket
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Ea.
Protection from moisture
NS 14/23