Learn More
DWK Life Sciences DURAN™ Expansion Adapter, socket/cone
Expansion from sleeve to core.
11.28 € - 836.00 €
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15931601
|
DWK Life Sciences
286524403 |
Socket NS 14/23, Cone NS 19/26 |
107.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10142021
|
DWK Life Sciences
286543203 |
Socket NS 24/29, Cone NS 45/40 | N/A | ||||||
15941601
|
DWK Life Sciences
286524806 |
Socket NS 14/23, Cone NS 29/32 | N/A | ||||||
15884808
|
DWK Life Sciences
286536002 |
Socket NS 19/26, Cone NS 24/29 |
134.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15894808
|
DWK Life Sciences
286536208 |
Socket NS 19/26, Cone NS 29/32 |
138.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15931611
|
DWK Life Sciences
286549403 |
Socket NS 29/32, Cone NS 45/40 |
254.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15834808
|
DWK Life Sciences
286524609 |
Socket NS 14/23, Cone NS 24/29 |
114.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15804828
|
DWK Life Sciences
286549009 |
Socket NS 29/32, Cone NS 60/46 |
651.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15814828
|
DWK Life Sciences
286549206 |
Socket NS 29/32, Cone NS 71/51 |
836.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
10161781
|
DWK Life Sciences
286542601 |
Socket NS 24/29, Cone NS 29/32 | N/A | ||||||
15921611
|
DWK Life Sciences
286548407 |
Socket NS 29/32, Cone NS 34/35 |
223.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10707101
|
DWK Life Sciences
286548407 |
Socket NS 29/32, Cone NS 34/35 | N/A | ||||||
15894818
|
DWK Life Sciences
286548604 |
Socket NS 29/32, Cone NS 50/42 |
379.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
Description
Adapters are used to connect two components with different NS sizes. DURAN™ expansion pieces allow the connection of a smaller sleeve to a larger core.
- Expansion pieces allow the connection of a smaller sleeve to a larger core and thus the use of differently sized ground joint components.
Specifications
10/Pk. |