Learn More
Karl Hecht™ DURAN™ Extraction Apparatus with Dimroth Condenser
This extraction apparatus is according to Soxhlet.
Brand: Karl Hecht™ 41151012
This item is not returnable.
View return policy
Description
- With standard grounds
- Flask joint size 29/32, condenser joint size of 34/35
- Joint sizes and capacity of extractors are durably marked
- Consists of extractor, dimroth condenser and round bottom flask
- Flask capacity of 100mL, extraction capacity of 70mL
Specifications
Soxhlet Extraction Apparatus | |
Borosilicate Glass 3.3 | |
29/32 |
100 mL (Flask), 70 mL (Extractor) | |
34/35 |