Learn More
Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet
For the extraction of liquids with specific light solvents. Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet are manufactured from durable DURAN™ tubing.
Brand:  Lenz Laborglasinstrumente™ 05395137
 View more versions of this product
Product Code. 10740612
  • 106.00 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc to DIN 12602 and Soxhlet
  • With cone NS 29/32
  • Available with/without NS stopcock
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Extractor Head
100 mL
Duran Glass
DIN 12602