Learn More
DWK Life Sciences DURAN™ Flask, Kjeldahl
For nitrogen determination.
Brand:  DWK Life Sciences 212315109
 View more versions of this product
Product Code. 11737981
  • 330.00 € / Pack of 10
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DURAN™ Kjeldahl flasks are subjected to extreme chemical and physical stresses. Therefore, the wall thickness should not be too thick to avoid overheating, which can lead to breakage due to remaining (permanent) stresses, on the one hand, and not too thin due to the noticeable glass removal in the alkaline medium, on the other hand. In order to offer the user the safest possible compromise while taking the technological conditions into account, the wall thickness of the DURAN™ Kjeldahl flasks in the body are 1-2 mm.
  • Wall thickness adapted to the application (glass removal by alkaline medium taken into account)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
115 mm
Borosilicate Glass (3.3)
10/Pk.
750 mL
For nitrogen determination
Autoclavable