Learn More
DWK Life Sciences DURAN™ Flat Flange Lid, 4 standard ground necks, with side necks: 2 x NS 29/32 angled; 1 x 14/23 parallel,, center neck: NS 29/32
The lid of a plane fan vessel is suitable for closing but also for combination with other ground joint components (e.g. dropping funnel, stirring shafts, etc.) due to the standard ground joints.
326.00 € - 348.00 €
Specifications
Material Borosilicate Glass (3.3)
For Use With (Application) Chemically and thermally demanding processes; Safe working under pressure or vacuum
Neck Style 2 x NS 29/32 angled; 1 x NS 14/23 parallel
No. per Pack 1/Pk.
Socket Joint NS 29/32
View More Specs
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10749101
View Documents
DWK Life Sciences
243924702
DN 100
326.00 €
Each
In Stock
to see stock.
Add to basket
10211811
View Documents
DWK Life Sciences
243925904
DN 150
348.00 €
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
10709681
View Documents
DWK Life Sciences
243925107
DN 120
334.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
Flat flange vessels are large volume reaction vessels which are regarded as an alternative to the round flasks and consist of a bottom part and a lid. These two parts are held together by means of a clamping ring. A seal or grinding grease prevent the grinding surfaces from tilting. The flat flange vessel can be combined with other components (e.g. dropping funnels, stirring shafts, ...) by means of various standard ground joints in the lid. Flat flange vessels are well suited for preparative work such as general reactions, distillations, evaporation and drying of solid and viscous substances. Due to their walls, flat flange vessels are suitable for safe working under pressure and vacuum.
  • With one middle neck: 29/32 and three side necks (standard ground): 3 x 29/32 parallel
  • Pressure and vacuum resistant due to the wall thickness and geometry
  • Tight closure due to standard ground joint
Specifications
Specifications
Borosilicate Glass (3.3)
2 x NS 29/32 angled; 1 x NS 14/23 parallel
NS 29/32
Chemically and thermally demanding processes; Safe working under pressure or vacuum
1/Pk.
Autoclavable