Learn More
DWK Life Sciences DURAN™ Flat sealing O-ring Gasket GL 45
Flat, flexible EPDM Rubber GL 45 sealing gasket forms an air tight seal between a DURAN™ GL 45 bottle and GL 45 connection caps.
Brand: DWK Life Sciences 1152752
9.29 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Soft flexible EPDM rubber forms an air-tight seal
- EPDM ensures an airtight seal with bottle
Specifications
40 mm ring gasket seal for GL45 multi-function caps. Circular EPDM Seals 1.5 mm thick with a 40 mm outer and 29 mm inner diameter. | |
GL 45 | |
EPDM |
Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer bewteen lab bottles; Supply of reagents to instruments | |
5/Pk. |