missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ GL 45 Hydra Square Bottle

Product Code. 16621076 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Ideal for the storage and delivery of high-performance liquid chromatography (HPLC) mobile phases and solvents.

  • Ideal for the storage and delivery of high-performance liquid chromatography (HPLC) mobile phases and solvents.
  • Increasing the number of GL 45 inlets on the bottles adds convenience: The five GL 45 necks on a purging bottle increases safety and reduces the risk of cross contamination and air intake.
  • Convenient – Purging bottle accepts up to 5 HPLC GL 45 cap
  • Safer – Capped bottles reduce the risk of volatile organic compounds (VOC) escape and spillage
  • Compatible - GL 45 threads fit standard HPLC connection caps
  • Manufactured from the material of choice for HPLC solvent bottles: borosilicate 3.3 glass offering a very low inorganic ion extractables profile
  • Clear DURAN™ is a borosilicate glass type 3.3 as specified by the international standard ISO 3585, and is classified as a ''Type I, Class A low-expansion borosilicate glass'' as defined by ASTM-E438-1992
  • 1 Liter Square space saving shape
  • Retrace Code, and certificate for manufacturing lot traceability
  • Durable white enamel volume graduations
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Cleaning Procedure Lab washer compatible
Color Clear
Diameter (Metric) Inner Neck 30 mm
For Use With (Application) HPLC Purge Bottle, Reagent Supply to Liquid Handling Systems and Analyzers
Includes 4 GL 45 Caps, 4 GL 45 Side Necks
Material Borosilicate Glass (3.3)
Length (Metric) 222.5 mm
Max. Temperature (Metric) 500°C
Quantity 1
Screw Color Blue
Screw Material Polypropylene
Sterility Non-sterile
Weight (Metric) 800 g
Thread Size (Metric) GL 45
Closure Color Blue
Graduations 100 to 800
Nominal Capacity 1000 mL
Certifications/Compliance DIN ISO 3585, Glass Type I according to the requirements of the current United States pharmacopoeia USP <660> Containers - Glass, European pharmacopoeia EP (3.2.1) Glass Containers for Pharmaceutical Use, Japanese pharmacopoeia JP <7.01> Test for Glass Containers for injections, This batch certificate confirms that ASTM Class A error limits required by USP are met. The certificate records the mean value, the standard deviation of the batch and the date of issue.
Closure Included Yes
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Narrow
Height (Metric) 222.5 mm
Closure Material Polypropylene
Diameter (Metric) 94 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.