missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Duran Glass Conical Centrifuge Tubes

Constructed from borosilicate glass. Lenz Laborglasinstrumente™ Duran Glass Conical Centrifuges Tubes are used to separate solids from liquids in chemical solutions.

Brand:  Lenz Laborglasinstrumente™ 05675000

 View more versions of this product

Product Code. 10253924

  • 56.00 € / Each
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Borosilicate glass
  • Conical bottom
Specifications

Specifications

ASTM D91
Borosilicate Glass
Cylindrical
Conical
Centrifuge Tube
100 mL
1
203 mm
Yes
700 x g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasinstrumente™ Duran Glass Conical Centrifuge Tubes > Capacity: 100mL; Length: 203mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.