Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Coil Condenser
Duran™ Glass Coil Condensers made of DURAN™ tubing.
Brand:  Lenz Laborglasinstrumente™ 05250204
 View more versions of this product
Product Code. 10003922
  • 132.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Duran™ Glass
Specifications
Specifications
DIN 12593 and in addition to DIN
For condensing vapors
1 Ea.
Duran Glass
Cooling Surface: 240 cm2
160 mm
NS 29/32
Coil Condenser
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Duran™ Glass Coil Condenser > Neck Size: 29/32, Jacket Length: 160 mm