Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Dimroth Condensers For Extractors
Brand:  Lenz Laborglasinstrumente™ 05246006
 View more versions of this product
Product Code. 10189510
  • 138.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Cooling Surface: 370cm2
1 Each
Duran Glass
For condensing vapors
45/40
Dimroth Condenser