Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Dimroth Condensers For Extractors
Brand:  Lenz Laborglasinstrumente™ 05246004
 View more versions of this product
Product Code. 10710622
  • 121.00 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Cooling Surface: 140cm2
1 Each
Duran Glass
For condensing vapors
29/32
Dimroth Condenser