Learn More
Lenz Laborglasinstrumente™ Double Walled Coil Condensers with GL Threads
Facilitates the condensing of vapors in the laboratory. Lenz Laborglasinstrumente™ Double Walled Coil Condensers with GL Threads are manufactured from durable DURAN™ tubing.
Brand:  Lenz Laborglasinstrumente™ 05250314
Product Code. 10121602
  • 106.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12593
  • With NS socket and cone
  • With cooling coil and jacket
  • Corrosion-free and resistant to thermal shock
  • Water connection: GL 14 threads (with two threaded plastic olives)
Specifications
Specifications
250 mm
1 Each
Duran Glass
Intensive Condenser