missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ DURAN™ Receiving Flask

Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ DURAN™ Receiving Flasks are manufactured from durable DURAN glass.

Brand:  Lenz Laborglasinstrumente™ 03063570

Product Code. 11816114

  • 68.00 € / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With spherical joint socket
  • With inscription label
  • Available in a range of capacities
Specifications

Specifications

1000 mL
Borosilicate Glass, Duran Glass
Rotary Evaporators
131 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasinstrumente™ DURAN™ Receiving Flask > Capacity: 1000mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.