Learn More
Lenz Laborglasinstrumente™ Reduction Adapters with Ground Joints
Provides for secure and easy connection from a large cone to a small socket in glass tubing. Lenz Laborglasinstrumente™ Reduction Adapters with Ground Joints are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 01501929
Product Code. 10111951
  • 12.00 € / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12257
  • High taper accuracy of the cones
  • Minimized need for grease
  • Excellent tightness
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
DIN 12257 and in addition to DIN standard
Duran Glass
1Each
NS 29/32
Reduction Adapter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Reduction Adapters with Ground Joints > Socket Size/Cone Size: NS 19/26/NS 29/32