Learn More
Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone
Provides for secure and easy leak-tight connection of apparatus in the laboratory. Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 01312529
This item is not returnable.
View return policy
Description
- Acc. to DIN 12257
- Available in a range of socket and thread sizes
- Minimized need for grease
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifications
1 Ea. | |
NS 29/32 | |
GL 25 | |
Duran Glass |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone > Socket Size: NS 29/32; Thread Size: GL 25
Spot an opportunity for improvement?Share a Content Correction